Learn More
Invitrogen™ LRRC28 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113971
Description
Immunogen sequence: KDEGLQYLER LYMKRNSLTS LPENLAQKLP NLVELYLHSN NIVVVPEAIG For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.
LRRC28 contains 11 LRR (leucine-rich) repeats. The function of the LRRC28 protein is not known.
Specifications
| LRRC28 | |
| Polyclonal | |
| Unconjugated | |
| LRRC28 | |
| 1300004K21Rik; 2210012C09Rik; 2310058O11Rik; leucine rich repeat containing 28; leucine-rich repeat-containing protein 28; LRRC28; UNQ436/PRO867 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 123355 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q86X40 | |
| LRRC28 | |
| Synthetic peptide directed towards the N terminal region of human LRRC28 (aa 36-85). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.