Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRRC28 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA558764
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558764 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558764 Supplier Thermo Scientific Supplier No. PA558764

Rabbit Polyclonal Antibody

Immunogen sequence: NIHLKGLPSY LYNKVIGCSG CGAPIQVSEV KLLSFSSGQR TVFLPAEVKA IGTEHDHVLP LQELAMRGLY HTYHSLLKDL NFL Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 81%.

LRRC28 contains 11 LRR (leucine-rich) repeats. The function of the LRRC28 protein is not known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRC28
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LRRC28
Gene Accession No. Q86X40
Gene Alias 1300004K21Rik; 2210012C09Rik; 2310058O11Rik; leucine rich repeat containing 28; leucine-rich repeat-containing protein 28; LRRC28; UNQ436/PRO867
Gene Symbols LRRC28
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LRRC28
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 123355
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.