Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRC29 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116268
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116268 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116268 Supplier Invitrogen™ Supplier No. PA5116268
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LRRC29 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRC29
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene LRRC29
Gene Accession No. Q8WV35
Gene Alias A630024J02Rik; FBL9; F-box and leucine-rich repeat protein 9; F-box protein FBL9; F-box/LRR-repeat protein 9; FBXL9; FBXL9P; leucine rich repeat containing 29; leucine-rich repeat-containing protein 29; LRRC29
Gene Symbols LRRC29
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 32-60 amino acids from the N-terminal region of human LRRC29.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26231
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.