Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRC3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547689
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547689 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547689 Supplier Invitrogen™ Supplier No. PA547689
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

In direct ELISAs, approximately 5% cross-reactivity with recombinant human LRRC4 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRC3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene LRRC3
Gene Accession No. Q9BY71
Gene Alias C21orf102; leucine rich repeat containing 3; leucine-rich repeat-containing protein 3; LRRC3; LRRC3A; UNQ9233/PRO31982
Gene Symbols LRRC3
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human LRRC3 Cys33-Asp204.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81543
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.