Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRRC30 Polyclonal Antibody, FITC, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS18377RFITC
This item is not returnable.
View return policy
Description
LRRC30 Polyclonal antibody specifically detects LRRC30 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)Specifications
| LRRC30 | |
| Polyclonal | |
| FITC | |
| LRRC30 | |
| leucine rich repeat containing 30; leucine-rich repeat-containing protein 30; LRRC30 | |
| KLH conjugated synthetic peptide derived from human LRRC30 (171-270). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300 | |
| A6NM36, Q3UV48 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 240131, 301711, 339291 | |
| -20° C, store in dark | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction