Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRC30 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559408
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559408 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559408 Supplier Invitrogen™ Supplier No. PA559408
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HNQLRVLPPE VGKLTRIVVL NLCGNRLKSL PREVSLLQCL KVLFVSMNCL TEVPAELSLC RKLEVLSLSH NCLSQ Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 88%.

LRRC30 (Leucine Rich Repeat Containing 30) is a Protein Coding gene. An important paralog of this gene is SHOC2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRC30
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LRRC30
Gene Accession No. A6NM36
Gene Alias leucine rich repeat containing 30; leucine-rich repeat-containing protein 30; LRRC30
Gene Symbols LRRC30
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LRRC30. Recombinant protein control fragment (Product #RP-98297).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 339291
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.