Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LRRC31 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144866
Description
Immunogen sequence: NVRFLKELIE LDISLRPSNF RDCGQWFRHL LYAVTKLPQI TEIGMKRWIL PASQEEELEC FDQDKKRSIH FDHGGF Antigen sequence identity: Mouse 71%; Rat 68%.

Specifications
LRRC31 | |
Polyclonal | |
Unconjugated | |
LRRC31 | |
E230002P03Rik; epididymis secretory protein Li 293; HEL-S-293; leucine rich repeat containing 31; leucine-rich repeat-containing protein 31; Lrrc31; UNQ9367/PRO34156 | |
Rabbit | |
Affinity chromatography | |
RUO | |
79782 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6UY01 | |
LRRC31 | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 476-551. Recombinant protein control fragment (Product #RP-109764). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction