Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRRC32/GARP Antibody (855151) [Janelia Fluor™ 646], Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB6055J
Description
LRRC32/GARP Monoclonal antibody specifically detects LRRC32/GARP in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
LRRC32/GARP | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2615 | |
Store at 4°C in the dark. | |
IgG2a |
Flow Cytometry, CyTOF | |
855151 | |
Flow Cytometry, CyTOF-ready | |
D11S833Eleucine-rich repeat-containing protein 32, GARPGarpin, Glycoprotein A repetitions predominantgarpin, leucine rich repeat containing 32 | |
recombinant human LRRC32/GARP, Accession # Q14392 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction