Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRRC32/GARP Antibody (GARP5) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP237710
Description
LRRC32/GARP Monoclonal specifically detects LRRC32/GARP in Human samples. It is validated for Western Blot.Specifications
LRRC32/GARP | |
Monoclonal | |
1 mg/mL | |
Western Blot 1-2 ug/ml | |
D11S833Eleucine-rich repeat-containing protein 32, GARPGarpin, Glycoprotein A repetitions predominantgarpin, leucine rich repeat containing 32 | |
Mouse | |
80 kDa | |
0.1 mg | |
Primary | |
Antibody recognizes GARP LRRC32, an approximately 80 kDa glycoprotein expressed e.g. on the surface of megakaryocytes, platelets and activated Treg cells. | |
Human | |
Purified |
Western Blot | |
GARP5 | |
Unconjugated | |
PBS (pH 7.4) with 15mM Sodium Azide | |
LRRC32 | |
Purified human sGARP protein. | |
Protein A purified | |
RUO | |
2615 | |
Protein A purified | |
Store at 4C. Do not freeze. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction