Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRRC35 Antibody (1B10), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259417AF750
This item is not returnable.
View return policy
Description
LRRC35 Monoclonal antibody specifically detects LRRC35 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
LRRC35 | |
Monoclonal | |
Alexa Fluor 750 | |
tubulin-specific chaperone cofactor E-like protein, El, LRRC35, tubulin folding cofactor E-like | |
Recombinant human LRRC35 (1-424aa) purified from E. coli | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, ELISA, Immunofluorescence | |
1B10 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
219899 | |
Store at 4°C in the dark. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction