Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRRC35 Antibody (1B10), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259417MFV610
Description
LRRC35 Monoclonal antibody specifically detects LRRC35 in Human samples. It is validated for Flow CytometrySpecifications
LRRC35 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
219899 | |
Store at 4C in the dark. | |
IgG2b κ |
Flow Cytometry | |
1B10 | |
Flow Cytometry | |
tubulin-specific chaperone cofactor E-like protein, El, LRRC35, tubulin folding cofactor E-like | |
Recombinant human LRRC35 (1-424aa) purified from E. coli | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction