Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRRC35 Antibody (OTI2A9), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272532H
Description
LRRC35 Monoclonal specifically detects LRRC35 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
LRRC35 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
TBCEL | |
Full length recombinant protein of human LRRC35 (NP_689928) produced in HEK293T cells. | |
0.1 mL | |
219899.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI2A9 | |
Western Blot, Immunohistochemistry | |
tubulin-specific chaperone cofactor E-like protein, El, LRRC35, tubulin folding cofactor E-like | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction