Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LRRC37A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560219
Description
Immunogen sequence: PGQPPEHHEV TVSPPGHHQT HHLASPSVSV KPPDVQLTIA AEPSAEVGTS LVHQEATTRL SGSGNDVEPP AIQHGGPPLL PESSEE Highest antigen sequence identity to the following orthologs: Mouse - 41%, Rat - 41%.
The function of this protein remains unknown.
Specifications
LRRC37A | |
Polyclonal | |
Unconjugated | |
LRRC37A | |
leucine rich repeat containing 37A; leucine-rich repeat-containing protein 37A; LRRC37; LRRC37A; LRRC37A1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9884 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.8 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A6NMS7 | |
LRRC37A | |
Recombinant protein corresponding to Human LRRC37A. Recombinant protein control fragment (Product #RP-104352). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction