Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRC37A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546525
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546525 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546525 Supplier Invitrogen™ Supplier No. PA546525
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: RESQDGLSSF GQPLWFKDLY KPLSATRINN HAWKLHKKSS NEDKILNRDP Sequence homology: Human: 100%.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRC37A
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene LRRC37A
Gene Accession No. A6NMS7
Gene Alias leucine rich repeat containing 37A; leucine-rich repeat-containing protein 37A; LRRC37; LRRC37A; LRRC37A1
Gene Symbols LRRC37A
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human LRRC37A (aa 1635-1684).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9884
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.