Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRC39 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585022
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585022 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585022 Supplier Invitrogen™ Supplier No. PA585022
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PEFIGRFQNL IVLDLSRNTI SEIPPGIGLL TRLQELILSY NKIKTVPKEL SNCASLEKLE LAVNRDICDL PQELSNLL Highest antigen sequence indentity to the following orthologs: Mouse - 92%, Rat - 94%.

Component of the sarcomeric M-band which plays a role in myocyte response to biomechanical stress. May regulate expression of other M-band proteins via an SRF-dependent pathway. Important for normal contractile function in heart.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRC39
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LRRC39
Gene Accession No. Q96DD0
Gene Alias densin hlg; leucine rich repeat containing 39; leucine-rich repeat-containing protein 39; LRRC39; Myomasp; Myosin-interacting M-band-associated stress-responsive protein; UNQ6500/PRO21368
Gene Symbols LRRC39
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LRRC39. Recombinant protein control fragment (Product #RP-108289).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 127495
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.