Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRRC4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | LRRC4 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
LRRC4 Polyclonal specifically detects LRRC4 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
LRRC4 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
64101 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
BAG, Brain Tumor Associated Protein LRRC4, Brain Tumor-Associated Protein BAG, Leucine Rich Repeat Containing 4, Leucine-Rich Repeat-Containing 4, Leucine-Rich Repeat-Containing Protein 4, Nasopharyngeal Carcinoma-Associated Gene 14 Protein, Netrin-G2 Ligand, NGL-2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 544-653 of human LRRC4 (NP_071426.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title