Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRRC4 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984367
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4367 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4367 Supplier Bioss Supplier No. BS1974R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LRRC4 protein function is currently unknown.

Specifications

Antigen LRRC4
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Lrrc4
Gene Accession No. Q9HBW1
Gene Alias BAG; brain tumor associated protein LRRC4; Brain tumor-associated protein BAG; Brain tumor-associated protein MBAG1; leucine rich repeat containing 4; leucine rich repeat containing 4 protein; leucine-rich repeat-containing 4; Leucine-rich repeat-containing protein 4; LIBG-like protein; LRRC4; LRRC4-like protein; MBAG1; NAG14; Nasopharyngeal carcinoma-associated gene 14 protein; Netrin-G2 ligand; NGL-2; NLG2; NLG-2; protein NGL-2; UNQ554/PRO1111
Gene Symbols Lrrc4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human LRRC4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64101
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.