Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LRRC40 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570275
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.
LRRC40 is a protein coding gene.
Specifications
LRRC40 | |
Polyclonal | |
Unconjugated | |
LRRC40 | |
2610040E16Rik; AI837674; dJ677H15.1; leucine rich repeat containing 40; leucine-rich repeat-containing protein 40; LOW QUALITY PROTEIN: leucine-rich repeat-containing protein 40; Lrrc40; testicular tissue protein Li 109 | |
Rabbit | |
Affinity chromatography | |
RUO | |
67144 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9CRC8 | |
LRRC40 | |
synthetic peptide directed towards the middle region of human LRRC40 (aa 513-562). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction