Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRC40 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598959
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598959 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598959 Supplier Invitrogen™ Supplier No. PA598959
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LRRC40 Polyclonal Antibody for Western Blot, ELISA

LRRC40 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRC40
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 4.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene LRRC40
Gene Accession No. Q9H9A6
Gene Alias 2610040E16Rik; AI837674; dJ677H15.1; leucine rich repeat containing 40; leucine-rich repeat-containing protein 40; LOW QUALITY PROTEIN: leucine-rich repeat-containing protein 40; Lrrc40; testicular tissue protein Li 109
Gene Symbols LRRC40
Host Species Rabbit
Immunogen Recombinant Human Leucine-rich repeat-containing protein 40 protein (1-602AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55631
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.