Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRC41 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA536596
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA536596 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA536596 Supplier Invitrogen™ Supplier No. PA536596
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 80 kDa. Purity is >95% by SDS-PAGE.

Probable substrate recognition component of an ECS (Elongin BC-CUL2/5-SOCS-box protein) E3 ubiquitin ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRC41
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide; pH 7.2
Gene LRRC41
Gene Accession No. Q15345, Q5M9H1, Q8K1C9
Gene Alias AA409966; AW555107; D630045E04Rik; D730026A16Rik; elongin BC-interacting leucine-rich repeat protein; leucine rich repeat containing 41; leucine-rich repeat-containing protein 41; Lrrc41; Muf1; MUF1 protein; MUF1 protein (MUF1); PP7759; Protein Muf1; RGD1311221; RP4-636H5.2
Gene Symbols LRRC41
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 402-453 of human LRRC41.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10489, 230654, 362566
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.