Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRC41 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562518
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562518 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562518 Supplier Invitrogen™ Supplier No. PA562518
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KPLKRFKRAA GKKGARTRQG PGAESEDLYD FVFIVAGEKE DGEEMEIGEV ACGALDGSDP SCLGLPALEA SQRF Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 96%.

Probable substrate recognition component of an ECS (Elongin BC-CUL2/5-SOCS-box protein) E3 ubiquitin ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRC41
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LRRC41
Gene Accession No. Q15345
Gene Alias AA409966; AW555107; D630045E04Rik; D730026A16Rik; elongin BC-interacting leucine-rich repeat protein; leucine rich repeat containing 41; leucine-rich repeat-containing protein 41; Lrrc41; Muf1; MUF1 protein; MUF1 protein (MUF1); PP7759; Protein Muf1; RGD1311221; RP4-636H5.2
Gene Symbols LRRC41
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LRRC41. Recombinant protein control fragment (Product #RP-99793).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10489
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.