Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRRC42 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | LRRC42 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
LRRC42 Polyclonal specifically detects LRRC42 in Human samples. It is validated for Western Blot.Specifications
| LRRC42 | |
| Polyclonal | |
| Rabbit | |
| Q9Y546 | |
| 115353 | |
| Synthetic peptides corresponding to LRRC42(leucine rich repeat containing 42) The peptide sequence was selected from the N terminal of LRRC42. Peptide sequence LIGFPEQIAEKLFSAAEARQKFTEPGAGLRALQKFTEAYGSLVLCSLCLR. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| dJ167A19.4, leucine rich repeat containing 42, leucine-rich repeat-containing protein 42, MGC8974 | |
| LRRC42 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title