Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LRRC42 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113544
Description
LRRC42 Polyclonal Antibody for Western Blot, IHC (P), ELISA
LRRC42 is a protein coding gene.
Specifications
| LRRC42 | |
| Polyclonal | |
| Unconjugated | |
| LRRC42 | |
| A930011F22Rik; AA536996; dJ167A19.4; leucine rich repeat containing 42; leucine-rich repeat-containing protein 42; LRRC42; MGC137941 protein; RGD1308919 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 115353 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9Y546 | |
| LRRC42 | |
| Recombinant Human Leucine-rich repeat-containing protein 42 protein (201-428AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction