Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRC42 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114760
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114760 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114760 Supplier Invitrogen™ Supplier No. PA5114760
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total LRRC42.

LRRC42 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRC42
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene LRRC42
Gene Accession No. Q4KM95, Q8R2U7, Q9Y546
Gene Alias A930011F22Rik; AA536996; dJ167A19.4; leucine rich repeat containing 42; leucine-rich repeat-containing protein 42; LRRC42; MGC137941 protein; RGD1308919
Gene Symbols LRRC42
Host Species Rabbit
Immunogen A synthesized peptide derived from human LRRC42(Accession Q9Y546), corresponding to amino acid residues S37-K87.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115353, 298309, 77809
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.