Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LRRC46 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110875
Description
Immunogen sequence: PTLTDLPLLP GVPMAGDSSP SATPAQGEET VPEAVSSPQA SSPTKKPCSL IPRGHQSSFW GRKGARAATA PKASVAEAPS TTKTT Highest antigen sequence indentity to the following orthologs: Mouse - 55%, Rat - 58%.
LRRC46 is a protein coding gene.
Specifications
| LRRC46 | |
| Polyclonal | |
| Unconjugated | |
| LRRC46 | |
| leucine rich repeat containing 46; leucine-rich repeat-containing protein 46; LRRC46 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 90506 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96FV0 | |
| LRRC46 | |
| Recombinant Protein corresonding to Human LRRC46. Recombinant protein control fragment (Product #RP-93283). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction