Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRC47 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552894
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552894 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552894 Supplier Invitrogen™ Supplier No. PA552894
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NCLDSFPAEL FRPGALPLLS ELAAADNCLR ELSPDIAHLA SLKTLDLSNN QLSEIPAELA DCPKLKEINF RGNKLRDKRL EKMVSGCQTR SILEYLRVGG RGGGKGKGRA E Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 92%.

LRRC47 gene ontology annotations related to this gene include phenylalanine-tRNA ligase activity; RNA binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRC47
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Lrrc47
Gene Accession No. Q505F5, Q8N1G4
Gene Alias 2900010D03Rik; AA960559; KIAA1185; leucine rich repeat containing 47; leucine-rich repeat-containing protein 47; LRRC47; mKIAA1185; RGD1308599; RP1-286D6.3
Gene Symbols Lrrc47
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LRRC47. Recombinant protein control fragment (Product #RP-88844).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 362672, 57470, 72946
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.