Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRC48 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546526
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546526 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546526 Supplier Invitrogen™ Supplier No. PA546526
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: NIFEYGLKQQ EKRKTELDTF SECVREAIQE NQEQGKRKIA KFEEKHLSLF Sequence homology: Cow: 93%; Dog: 86%; Guinea Pig: 85%; Horse: 93%; Human: 100%; Rabbit: 92%; Rat: 86%.

Component of the nexin-dynein regulatory complex (N-DRC) a key regulator of ciliary/flagellar motility which maintains the alignment and integrity of the distal axoneme and regulates microtubule sliding in motile axonemes. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRC48
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene DRC3
Gene Accession No. Q9H069
Gene Alias CFAP134; DRC3; Dynein regulatory complex subunit 3; dynein regulatory complex subunit 3; LOW QUALITY PROTEIN: dynein regulatory complex subunit 3; leucine rich repeat containing 48; leucine-rich repeat-containing protein 48; Lrrc48; RGD1309150
Gene Symbols DRC3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human LRRC48.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83450
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.