Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRC49 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553332
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553332 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553332 Supplier Invitrogen™ Supplier No. PA553332
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SHVDNLNGLD SLTELNLRHN QITFVRDVDN LPCLQHLFLS FNNISSFDSV SCLADSSSLS DITFDGNPIA QESWYKHTVL QNMMQLRQLD MKRITEEERR MASVLAKKEE EKKRESHKQS LLKEKKRLTI NNVARQWDLQ QQRVA Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.

LRRC49 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRC49
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LRRC49
Gene Accession No. Q8IUZ0
Gene Alias AI430817; AW050057; D430025H09Rik; leucine rich repeat containing 49; leucine-rich repeat-containing protein 49; Lrrc49; p79; PGs4; RGD1309466; Tubulin polyglutamylase complex subunit 4
Gene Symbols LRRC49
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LRRC49. Recombinant protein control fragment (Product #RP-90789).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54839
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.