Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRRC4B/NGL3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981094
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1094 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1094 Supplier Bioss Supplier No. BS11090R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen LRRC4B/NGL3
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene LRRC4B
Gene Accession No. Q9NT99
Gene Alias A830007M12; HSM; leucine rich repeat containing 4B; Leucine-rich repeat-containing protein 4B; leucine-rich repeats and immunoglobulin-like domains 4; Lrig4; LRRC4B; Netrin-G3 ligand; NGL-3; protein NGL-3
Gene Symbols LRRC4B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human LRRC4B/NGL3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 94030
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.