Learn More
Invitrogen™ LRRC4C Monoclonal Antibody (1G5)

Description
Peptide Sequence: AVMLVIFYKM RKQHHRQNHH APTRTVEIIN VDDEITGDTP MESHLPMPAI EHEHLNHYNS YKSPFNHTTT VNTINSIHSS VHEPLLIRMN SKDNVQETQI.
Specifications
Specifications
| Antigen | LRRC4C |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 1G5 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | LRRC4C |
| Gene Accession No. | Q9HCJ2 |
| Gene Alias | 6430556C10Rik; KIAA1580; leucine rich repeat containing 4C; leucine-rich repeat-containing protein 4C; Lrrc4c; Netrin-G1 ligand; NGL1; NGL-1; RGD1311013; UNQ292/PRO331 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.