Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRC4C Monoclonal Antibody (755720)
GREENER_CHOICE

Catalog No. PIMA524363
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524363 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524363 Supplier Invitrogen™ Supplier No. MA524363
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

In direct ELISAs, no cross-reactivity with recombinant human (rh) LRRC4 or rhNGL-3/LRRC4B is observed. Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.

Leucine-rich repeat-containing 4C (LRRC4C), also known as Netrin-G ligand 2 (NGL-2), is a gene highlighted for its expression in the nervous system, particularly the brain. It is a member of the leucine-rich repeat superfamily, which plays a significant role in cell adhesion and signaling pathways critical for synaptic connectivity and neuronal development. LRRC4C is involved in neural processes such as axon guidance and synapse formation, aiding in the establishment of functional neural circuits. Additionally, research has suggested that LRRC4C may function as a tumor suppressor, particularly in gliomas, indicating its potential role in neuro-oncological disorders. Variations or deletions in this gene have been linked to neurodevelopmental disorders, demonstrating its importance in normal brain function and development. Furthermore, LRRC4C's expression patterns and associations with cancer also suggest its potential as a prognostic marker in clinical settings.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRC4C
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 755720
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene LRRC4C
Gene Accession No. Q9HCJ2
Gene Alias 6430556C10Rik; KIAA1580; leucine rich repeat containing 4C; leucine-rich repeat-containing protein 4C; Lrrc4c; Netrin-G1 ligand; NGL1; NGL-1; RGD1311013; UNQ292/PRO331
Gene Symbols LRRC4C
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human NGL-1/LRRC4C Gln45-Lys527 (predicted).
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57689
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.