Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRC4C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590507
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590507 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590507 Supplier Invitrogen™ Supplier No. PA590507
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PFSYFSTVTV ETMEPSQDEA RTTDNNVGPT PVVDWETTNV TTSLTPQSTR STEKTFTIPV TDINSGIPGI D; Positive Samples: Rat eye; Cellular Location: Cell junction, Single-pass type I membrane protein, postsynaptic cell membrane, synapse.

Leucine-rich repeat-containing 4C (LRRC4C), also known as Netrin-G ligand 2 (NGL-2), is a gene highlighted for its expression in the nervous system, particularly the brain. It is a member of the leucine-rich repeat superfamily, which plays a significant role in cell adhesion and signaling pathways critical for synaptic connectivity and neuronal development. LRRC4C is involved in neural processes such as axon guidance and synapse formation, aiding in the establishment of functional neural circuits. Additionally, research has suggested that LRRC4C may function as a tumor suppressor, particularly in gliomas, indicating its potential role in neuro-oncological disorders. Variations or deletions in this gene have been linked to neurodevelopmental disorders, demonstrating its importance in normal brain function and development. Furthermore, LRRC4C's expression patterns and associations with cancer also suggest its potential as a prognostic marker in clinical settings.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRC4C
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.01 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene LRRC4C
Gene Alias 6430556C10Rik; KIAA1580; leucine rich repeat containing 4C; leucine-rich repeat-containing protein 4C; Lrrc4c; Netrin-G1 ligand; NGL1; NGL-1; RGD1311013; UNQ292/PRO331
Gene Symbols LRRC4C
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 450-520 of human LRRC4C (NP_0659801).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 311236
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.