Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRRC50 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP217171
Description
LRRC50 Polyclonal specifically detects LRRC50 in Mouse samples. It is validated for Western Blot.Specifications
LRRC50 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
CILD13, DKFZp434A119, FLJ25330, leucine rich repeat containing 50, ODA7leucine-rich repeat-containing protein 50, outer row dynein assembly 7 homolog | |
Rabbit | |
80 kDa | |
0.1 mL | |
Primary | |
Mouse. Species reactivity based on sequence identity: Human (100%), Rat (86%), Bovine (88%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot | |
0.96mg/mL | |
Western Blot 1:500-1:3000 | |
Q8NEP3 | |
DNAAF1 | |
Recombinant protein encompassing a sequence within the N-terminus region of human LRRC50. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
123872 | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction