Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRRC50 Antibody (OTI2C4), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272536FR
Description
LRRC50 Monoclonal specifically detects LRRC50 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry.Specifications
LRRC50 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
DNAAF1 | |
Full length recombinant protein of human LRRC50 (NP_848547) produced in HEK293T cells. | |
0.1 mL | |
123872.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry | |
OTI2C4 | |
Western Blot, Flow Cytometry | |
CILD13, DKFZp434A119, FLJ25330, leucine rich repeat containing 50, ODA7leucine-rich repeat-containing protein 50, outer row dynein assembly 7 homolog | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat, Canine, Primate, Monkey | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction