Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRRC50 Antibody (OTI2C4), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272536MFV500
Description
LRRC50 Monoclonal antibody specifically detects LRRC50 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Flow CytometrySpecifications
LRRC50 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Canine, Primate, Monkey | |
Purified |
Flow Cytometry | |
OTI2C4 | |
Flow Cytometry | |
CILD13, DKFZp434A119, FLJ25330, leucine rich repeat containing 50, ODA7leucine-rich repeat-containing protein 50, outer row dynein assembly 7 homolog | |
Full length human recombinant protein of human LRRC50(NP_848547) produced in HEK293T cell. | |
0.1 mL | |
Cell Biology, Cytoskeleton Markers, Signal Transduction | |
123872 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction