Learn More
Invitrogen™ LRRC50 Monoclonal Antibody (OTI2C4)

Description
LRRC50 Monoclonal Antibody for Western Blot, Flow
Specifications
Specifications
| Antigen | LRRC50 |
| Applications | Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Clone | OTI2C4 |
| Concentration | 2.74 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | DNAAF1 |
| Gene Accession No. | Q6AYH9, Q8NEP3, Q9D2H9 |
| Gene Alias | 4930457P18Rik; CILD13; Dnaaf1; dynein (axonemal) assembly factor 1; dynein assembly factor 1, axonemal; dynein axonemal assembly factor 1; dynein, axonemal assembly factor 1; dynein, axonemal, assembly factor 1; hypothetical protein LOC386722; hypothetical protein LOC523187; leucine rich repeat containing 50; leucine-rich repeat-containing protein 50; LLRC50; LOW QUALITY PROTEIN: dynein assembly factor 1, axonemal; lrrc50; ODA7; outer row dynein assembly 7 homolog; RGD1310542; switch hitter; swt; testicular tissue protein Li 110; unm tm317b; zgc:56169 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.