Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRRC50 Monoclonal Antibody (OTI2C4), TrueMAB™, OriGene
SDP

Catalog No. 502540262
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0262 30 μL
50-167-5204 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0262 Supplier OriGene Supplier No. TA504546S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LRRC50 Monoclonal antibody specifically detects LRRC50 in Human samples. It is validated for Flow Cytometry, Western Blot

Specifications

Antigen LRRC50
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI2C4
Concentration 2.74 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene NANP
Gene Accession No. Q8TBE9
Gene Alias 4930457P18Rik; CILD13; Dnaaf1; dynein (axonemal) assembly factor 1; dynein assembly factor 1, axonemal; dynein axonemal assembly factor 1; dynein, axonemal assembly factor 1; dynein, axonemal, assembly factor 1; hypothetical protein LOC386722; hypothetical protein LOC523187; leucine rich repeat containing 50; leucine-rich repeat-containing protein 50; LLRC50; LOW QUALITY PROTEIN: dynein assembly factor 1, axonemal; lrrc50; ODA7; outer row dynein assembly 7 homolog; RGD1310542; switch hitter; swt; testicular tissue protein Li 110; unm tm317b; zgc:56169
Host Species Mouse
Immunogen Full length human recombinant protein of human NANP produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140838
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.