Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRC50 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109642
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109642 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109642 Supplier Invitrogen™ Supplier No. PA5109642
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VATEGVFVTE LDGTRTEDLE TIRLETKETF CIDDLPDLED DDETGKSLED QNMCFPKIEV ISSLSDDSDP ELDYTSLPVL ENLPTDTLSN IFAVSKDTSK AARVPFTDIF KKEAKRDLEI RKQDTKSPRP LIQELSDEDP SGQLLMPPTC QRDAAPLTSS GDRDSDFLAA SSPVPTESAA TPPETCVGVA QPSQALPTWD LTAFPAPKAS.

The protein encoded by this gene is cilium-specific and is required for the stability of the ciliary architecture. It is involved in the regulation of microtubule-based cilia and actin-based brush border microvilli. Mutations in this gene are associated with primary ciliary dyskinesia-13.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRC50
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.31 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene DNAAF1
Gene Accession No. Q6AYH9
Gene Alias 4930457P18Rik; CILD13; Dnaaf1; dynein (axonemal) assembly factor 1; dynein assembly factor 1, axonemal; dynein axonemal assembly factor 1; dynein, axonemal assembly factor 1; dynein, axonemal, assembly factor 1; hypothetical protein LOC386722; hypothetical protein LOC523187; leucine rich repeat containing 50; leucine-rich repeat-containing protein 50; LLRC50; LOW QUALITY PROTEIN: dynein assembly factor 1, axonemal; lrrc50; ODA7; outer row dynein assembly 7 homolog; RGD1310542; switch hitter; swt; testicular tissue protein Li 110; unm tm317b; zgc:56169
Gene Symbols DNAAF1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 516-725 of human DNAAF1 (NP_848547.4).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361419
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.