Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRRC52 Antibody (1037552), Allophycocyanin/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB10260APCCY7
Description
LRRC52 Monoclonal antibody specifically detects LRRC52 in Human samples. It is validated for Flow CytometrySpecifications
LRRC52 | |
Monoclonal | |
APC-Cyanine7 | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
440699 | |
Store at 4C in the dark. Do not freeze. | |
IgG1 |
Flow Cytometry | |
1037552 | |
Flow Cytometry | |
FLJ25811, leucine rich repeat containing 52, leucine-rich repeat-containing protein 52 | |
Human embryonic kidney cell HEK293-derived human LRRC52, Ser24-Asp244, Accession # Q8N7C0 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction