Learn More
LRRC52 Antibody (1037552), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB10260MFV500
Description
LRRC52 Monoclonal antibody specifically detects LRRC52 in Human samples. It is validated for Flow CytometrySpecifications
LRRC52 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Flow Cytometry | |
1037552 | |
Flow Cytometry | |
FLJ25811, leucine rich repeat containing 52, leucine-rich repeat-containing protein 52 | |
Human embryonic kidney cell HEK293-derived human LRRC52, Ser24-Asp244, Accession # Q8N7C0 | |
RUO | |
440699 | |
Store at 4C in the dark. |