Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRC52 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5107159
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5107159 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5107159 Supplier Invitrogen™ Supplier No. PA5107159
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Sequence homology: Cow: 92%; Dog: 75%; Guinea Pig: 83%; Horse: 75%; Human: 100%; Mouse: 83%; Pig: 92%; Rabbit: 93%; Rat: 92%.

Auxiliary protein of the large-conductance, voltage and calcium-activated potassium channel (BK alpha). Modulates gating properties by producing a marked shift in the BK channel's voltage dependence of activation in the hyperpolarizing direction, and in the absence of calcium. KCNU1 channel auxiliary protein. May modulate KCNU1 gating properties. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRC52
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Gene LRRC52
Gene Accession No. Q8N7C0
Gene Alias 4930413P14Rik; BK channel auxiliary gamma subunit LRRC52; BK channel auxilliary gamma subunit LRRC52; leucine rich repeat containing 52; leucine-rich repeat-containing protein 52; LRRC52
Gene Symbols LRRC52
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal region of human LRRC52.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 440699
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.