Learn More
Invitrogen™ LRRC52 Polyclonal Antibody

Description
LRRC52 Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | LRRC52 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | LRRC52 |
| Gene Accession No. | Q8N7C0 |
| Gene Alias | 4930413P14Rik; BK channel auxiliary gamma subunit LRRC52; BK channel auxilliary gamma subunit LRRC52; leucine rich repeat containing 52; leucine-rich repeat-containing protein 52; LRRC52 |
| Gene Symbols | LRRC52 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.