Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LRRC57 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550443
Description
The antibody detects endogenous levels of total LRRC57 protein.
LRRC57 contains 7 LRR (leucine rich repeats). However, the exact function remains unknown.
Specifications
LRRC57 | |
Polyclonal | |
Unconjugated | |
LRRC57 | |
2810002D13Rik; AA407405; DKFZp686H1865; FLJ36812; leucine rich repeat containing 57; leucine-rich repeat-containing protein 57; LRC57; LRRC57 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
255252 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1.7 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q8N9N7 | |
LRRC57 | |
Full length fusion protein of Human LRRC57. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction