Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRRC59 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306264JF646
Description
LRRC59 Polyclonal antibody specifically detects LRRC59 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
LRRC59 | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
leucine rich repeat containing 59 | |
Produced in rabbits immunized with E. coli-derived Human LRRC59 fragment. | |
0.1 mL | |
Signal Transduction | |
55379 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunoprecipitation | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction