Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRC59 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543327
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543327 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543327 Supplier Invitrogen™ Supplier No. PA543327
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: KEYDALKAAK REQEKKPKKE ANQAPKSKSG SRPRKPPPRK HTRSWAVLKL Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.

Required for nuclear import of FGF1, but not that of FGF2. Might regulate nuclear import of exogenous FGF1 by facilitating interaction with the nuclear import machinery and by transporting cytosolic FGF1 to, and possibly through, the nuclear pores. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRC59
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Lrrc59
Gene Accession No. Q96AG4
Gene Alias AA959742; C78668; hypothetical protein PRO1855; leucine rich repeat containing 59; leucine-rich repeat-containing protein 59; Leucine-rich repeat-containing protein 59, N-terminally processed; leucine-rich repeat-containing protein 59; LOW QUALITY PROTEIN: leucine-rich repeat-containing protein 59; LRRC59; p34; PRO1855; Protein p34; Rbp34; Ribosome-binding protein p34
Gene Symbols Lrrc59
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human LRRC59.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55379
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.