Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LRRC59 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583160
Description
Immunogen sequence: CRVTELQQQP LCTSVNTIYD NAVQGLRRHE ILQWVLQTDS QQ.
Required for nuclear import of FGF1, but not that of FGF2. Might regulate nuclear import of exogenous FGF1 by facilitating interaction with the nuclear import machinery and by transporting cytosolic FGF1 to, and possibly through, the nuclear pores. [UniProt].
Specifications
LRRC59 | |
Polyclonal | |
Unconjugated | |
Lrrc59 | |
AA959742; C78668; hypothetical protein PRO1855; leucine rich repeat containing 59; leucine-rich repeat-containing protein 59; Leucine-rich repeat-containing protein 59, N-terminally processed; leucine-rich repeat-containing protein 59; LOW QUALITY PROTEIN: leucine-rich repeat-containing protein 59; LRRC59; p34; PRO1855; Protein p34; Rbp34; Ribosome-binding protein p34 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
287633, 55379, 98238 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5RJR8, Q922Q8, Q96AG4 | |
Lrrc59 | |
Recombinant protein corresponding to Human LRRC59. Recombinant protein control fragment (Product #RP-95601). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction