Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRC59 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585486
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585486 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585486 Supplier Invitrogen™ Supplier No. PA585486
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Keep as concentrated solution. Predicted reactivity: Mouse (93%), Rat (92%), Pig (96%), Bovine (95%). Positive Control: 293T, A431, HeLa, HepG2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Required for nuclear import of FGF1, but not that of FGF2. Might regulate nuclear import of exogenous FGF1 by facilitating interaction with the nuclear import machinery and by transporting cytosolic FGF1 to, and possibly through, the nuclear pores. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRC59
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene Lrrc59
Gene Accession No. Q922Q8, Q96AG4
Gene Alias AA959742; C78668; hypothetical protein PRO1855; leucine rich repeat containing 59; leucine-rich repeat-containing protein 59; Leucine-rich repeat-containing protein 59, N-terminally processed; leucine-rich repeat-containing protein 59; LOW QUALITY PROTEIN: leucine-rich repeat-containing protein 59; LRRC59; p34; PRO1855; Protein p34; Rbp34; Ribosome-binding protein p34
Gene Symbols Lrrc59
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human LRRC59. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55379, 98238
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.