Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRRC59 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988497
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8497 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8497 Supplier Bioss Supplier No. BS8361R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

C3 is a key component of the complement system since classical and alternative activation pathways merge at the C3 activation step when C3 is split into C3a and C3b. The molecular mass of C3 is 185 kDa and it consists of two chains (110 kDa and 75 kDa) held together by disulfide bonds.

Specifications

Antigen LRRC59
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Lrrc59
Gene Accession No. Q96AG4
Gene Alias AA959742; BOS_18977; C78668; leucine rich repeat containing 59; leucine rich repeat containing 59-like; Leucine-rich repeat-containing protein 59; Leucine-rich repeat-containing protein 59, N-terminally processed; LOC100341121; Lrrc59; p34; PRO1855; Protein p34; Rbp34; Ribosome-binding protein p34
Gene Symbols Lrrc59
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human LRRC59.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55379
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.