Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRRC6 Antibody (OTI7E9), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272542B
Description
LRRC6 Monoclonal specifically detects LRRC6 in Human samples. It is validated for Immunohistochemistry.Specifications
LRRC6 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
LRRC6 | |
Full length recombinant protein of human LRRC6 (NP_036604) produced in HEK293T cells. | |
0.1 mL | |
23639.0 | |
Store at 4°C in the dark. |
Immunohistochemistry | |
OTI7E9 | |
Immunohistochemistry | |
leucine rich repeat containing 6, Leucine-rich testis-specific protein, LRTPleucine-rich repeat-containing protein 6, Testis-specific leucine-rich repeat protein, TSLRPtestis specific leucine rich repeat protein | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction