Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRRC6 Monoclonal Antibody (OTI6D4), TrueMAB™, OriGene
SDP

Catalog No. 501680972
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-0972 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-0972 Supplier OriGene Supplier No. TA806108
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LRRC6 may be involved in spermatocytogenesis or prophase of meiosis.

Specifications

Antigen LRRC6
Applications Western Blot
Classification Monoclonal
Clone OTI6D4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene LRRC6
Gene Accession No. Q86X45
Gene Alias CILD19, LRTP, TSLRP
Gene Symbols LRRC6
Host Species Mouse
Immunogen Full length human recombit protein of human LRRC6 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23639
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.